missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Viral/Pathogen Ultra Enzyme Mix

Catalog No. A42366
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A42366 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A42366 Supplier Applied Biosystems™ Supplier No. A42366

This enzyme mix is included in the MagMAX Viral/Pathogen Nucleic Acid Ultra Isolation Kit.

This enzyme mix is included in the MagMAX Viral/Pathogen Nucleic Acid Ultra Isolation Kit (Cat. No. A42356). It is made available separately for applications that require more enzyme mix than is provided in the kit.

Specifications

Content And Storage Store in freezer (-5° to -30°C).
Sample Type Liquid Samples (e.g. Serum), Viral Samples, Buccal Samples, Blood, Cells, Bacteria, Fungi, Yeast
Efficiency High
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
Product Line MagMAX
Product Type Viral/Pathogen Ultra Enzyme Mix
Quantity 50 mL
Shipping Condition Dry Ice
Target Viral DNA/RNA, Pathogen DNA/RNA, Total Nucleic Acids
Volume (Metric) 50 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.