missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Viral/Pathogen II (MVP II) Nucleic Acid Isolation Kit

Catalog No. A48383
Encompass
Change view
Click to view available options
Quantity:
Up to 2000 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A48383 Up to 2000 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A48383 Supplier Applied Biosystems™ Supplier No. A48383

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The MagMAX™ Viral/Pathogen II (MVP II) Nucleic Acid Isolation Kit is specifically designed to recover RNA and DNA from viral particles contained in viral transport media (VTM). The kit utilizes MagMAX™ magnetic-bead technology, ensuring reproducible recovery of high-quality nucleic acid.

  • Compatible with the TaqPath COVID-19 Combo Kit (Cat. No. A47814)
  • Automation-ready protocols designed for the KingFisher Flex system
  • Fast procedure allows for 96 samples to be processed in less than 30 minutes
  • Flexible protocol accommodates sample volume inputs from 200 to 400 μL of transfer medium
  • No need for carrier RNA
  • Elution volume of 50 μL

Specifications

Content And Storage Now available in a convenient single-box format with a single expiration date.

• 550 mL Binding Solution
• 1,000 mL Wash Solution
• 20 mL MVP II Binding Beads
• 10 mL Proteinase K
• 100 mL Elution Buffer

Store at room temperature.

Format 96 Deep-well Plate
Purification Time 30 min.
Sample Type Nasopharyngeal Swabs and Saliva
High-throughput Compatibility High Throughput-Compatible
For Use With (Application) Real-Time PCR
No. of Reactions 2000 Preps
Product Line MagMAX
Quantity Up to 2000 preps
Shipping Condition Room Temperature
Target Viral DNA/RNA
Show More Show Less
How do I download the Pathogen Interpretive Software?

Once you place your order, please contact our Instrument Service team at 1-800-955-6288 option 3, 1 (this kit is only available in North America). You will be given an access code to the TaqPath Monkeypox/Orthopox Virus DNA Kit eLearning where you’ll learn how to install, update, and download the Pathogen Interpretive Software package for the TaqPath Monkeypox/Orthopox Virus DNA Kit.

With the TaqPath Monkeypox/Orthopox Virus DNA Kit, what software should I use for data analysis?

Results are analyzed using the following software:
- Applied Biosystems Pathogen Interpretive Software v1.1
- SAE Administrator Console Dx v1.0 or v1.2 (for security and audit functions)
- Pathogen Interpretive Software 1.0.0 DAT (SAE Profile)

Please use the appropriate assay panel and template for your instrument:
- MPXOPX-EUA_QS5-9601_1.0.0.zip
- MPXOPX-EUA_QS5Dx-9602_1.0.0.zip
- MPXOPX-EUA_QS7F-384_1.0.0.zip

Note: These zip files are located in EducationConnect (www.thermofisher.com/educationconnect). The eLearning course needs to be completed to be granted access.

What is the instrument software configuration required to use the TaqPath Monkeypox/Orthopox Virus DNA Kit?

To obtain the correct analysis software or firmware version for your real-time PCR instrument, go to thermofisher.com/validatedfirmware, then select your instrument in the Real-Time PCR section.

Which instruments are authorized under the EUA for use with the TaqPath Monkeypox/Orthopox Virus DNA Kit?

Here are the instruments that can be used with the TaqPath Monkeypox/Orthopox Virus DNA Kit:
- Applied Biosystems QuantStudio 5 Real-Time PCR System (96-well, 0.1 mL block)
- Applied Biosystems QuantStudio 5 Dx Real-Time PCR System (96-well, 0.2 mL block)
- Applied Biosystems QuantStudio 7 Flex Real-Time PCR System (384-well block)

I would like to use the TaqPath Monkeypox/Orthopox Virus DNA Kit. Which extraction kit should I use to extract DNA from the sample?

Performance was established using the MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit (Cat. No. A48383) in combination with the KingFisher Flex Magnetic Particle Processor with 96 Deep-Well Head and the KingFisher Flex 96 Deep-Well Heating Block (MVP_2Wash_400_Flex program). Other purification methods may lead to deviations in the expected result.

Which specimen types are compatible with the TaqPath Monkeypox/Orthopox Virus DNA Kit?

The TaqPath Monkeypox/Orthopox Virus DNA Kit protocol is currently authorized for use with lesion swab specimens in universal transport media (UTM) or viral transport media (VTM) from individuals suspected of monkeypox infection by their healthcare provider.

What instruments, consumables, and reagents do I need for the TaqPath Monkeypox/Orthopox Virus DNA Kit? Where can I find the full list of EUA products and protocols?

Go to www.thermofisher.com/monkeypox and download the Instructions for Use (IFU) to see the workflow outlining the instruments, consumables, and reagents required to run Monkeypox detection experiments.

What are the reporter dyes and targets for the TaqPath Monkeypox/Orthopox Virus DNA Kit?

The reporter dyes used are as follows:
- Monkeypox virus: FAM dye
- Orthopoxvirus: VIC dye
- RNase P: JUN dye

The TaqPath Monkeypox/Orthopox Virus DNA Kit probes contain QSY and MGB quenchers, which do not fluoresce. Select None for Quencher in the instrument set up procedure.

What are the components of the TaqPath Monkeypox/Orthopox Virus DNA Kit? Is the master mix included, or does it need to be purchased separately? Are there any other required materials that are not supplied?

The TaqPath Monkeypox/Orthopox Virus DNA Kit includes the following components:
- TaqPath Monkeypox/Orthopox Virus Multiplex Assay: Multiplexed real-time PCR assay containing primers/probes specific to monkeypox virus, non-variola orthopoxvirus targets, and human RNase P.
- TaqPath Monkeypox/Orthopox Virus DNA Positive Control: Positive DNA control that contains templates specific to monkeypox virus, non-variola orthopoxvirus targets, and human RNase P regions targeted by the assay.

Note:
- The master mix is not included in the kit and needs to be purchased separately: TaqPath BactoPure Microbial Detection Master Mix (No ROX) (Cat. No. A52703, A52704, or A52705)
- TE Buffer (Cat. No. 12090015) and Nuclease-free water (MLS) are required and not supplied with the kit.

Do you offer the Proteinase K that is included in the MagMAX Viral/Pathogen II (MVP II) Nucleic Acid Isolation Kit (Cat. No. A48383) as a stand-alone product?

No standalone components are validated for In Vitro Diagnostic (IVD) use with this kit.


For In Vitro Diagnostic Use.