missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Plant RNA Isolation Kit

Catalog No. A47157
Encompass
Change view
Click to view available options
Quantity:
1000 Preps
384 Preps
96 Preps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A47157 1000 Preps
A33784 96 Preps
A33899 384 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A47157 Supplier Applied Biosystems™ Supplier No. A47157
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

The MagMAX Plant RNA Isolation Kit is designed for automated high-throughput or manual purification of total RNA from wide variety of plant species and tissue types.

The MagMAX Plant RNA Isolation Kit is designed for automated high-throughput or manual purification of total RNA from wide variety of plant species and tissue types. It utilizes paramagnetic bead technology, enabling high yields and robust performance. The high binding capacity, uniform particle size, and rapid magnetic response of MagMAX magnetic beads makes the technology ideal for high-throughput automatic nucleic acid purification, as well as for manual nucleic acid purification by low- throughput users. The resulting high quality purified RNA is free of proteins, nucleases, and other contaminants or inhibitors, and can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, and other enzymatic reactions.

Versatile—suitable for any plant type and part, including woody, polyphenol-rich, and lignified samples
High yields of pure total RNA
High quality—isolates intact total RNA, free of inhibitors and gDNA contamination. Isolated RNA is suitable for long term storage at -20°C.
Flexible—easily adjustable for high-throughput automation platforms and manual processing

Applications
The MagMAX Plant RNA Isolation Kit is designed for fast extraction of high purity RNA suitable for all conventional molecular biology applications, including:
• RT-PCR
• RT-qPCR
• Northern blotting
• Nuclease protection assay

Benefits of MagMAX magnetic bead–based purification
The magnetic bead–based purification format of the kit allows for easy scaling of the number of samples processed either manually with a magnetic stand or with a magnetic particle processor such as the KingFisher Duo Prime or KingFisher Flex processors.

Magnetic beads offer many benefits compared to other technologies for isolating nucleic acids. Beads bind the RNA and DNA more efficiently than glass fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of undigested tissue particulates clogging these items during the extraction process. This clogging issue is of particular concern with fibrous plant tissue samples that that are not fully lysed during the disruption and homogenization steps.

Optimized bead surface chemistry
The Dynabeads MyOne Carboxylic Acid utilized in the MagMAX Plant RNA Isolation Kit has an optimized surface chemistry with high specific surface area that allows efficient kinetics and high sensitivity in nucleic acid capture. These aspects make Dynabeads magnetic technology ideal for the recovery of the RNA targets found in a variety of plant samples. Once captured, the RNA is then eluted in 100 μL and ready for downstream applications.

Specifications

Content And Storage • 750 mL Lysis Buffer; room temperature
• 30 mL RNA Binding Beads; 4°C
• 3.4 mL DNase I; –20°C
• 10.5 mL DNase I Reconstitution Buffer; –20°C
• 32 mL Manganese Chloride Solution; –20°C
• 125 mL DNase I Buffer (2X); –20°C
• 1146 mL Wash Solution 1 (concentrated); room temperature
• 521 mL Wash Solution 2 (concentrated); room temperature
• 210 mL Rebinding Buffer; room temperature
• 313 mL Nuclease-free Water; room temperature

Isolation Technology Magnetic Bead
Purification Time Less than 1 hr. when using automated protocol
Elution Volume 100 μL
Sample Type Plant Samples
Final Product Type Total RNA
High-throughput Compatibility High-throughput Compatible
For Use With (Equipment) KingFisher™ Duo Prime, KingFisher™ Flex, KingFisher™ Apex
For Use With (Application) RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling
Quantity 1000 Preps
Shipping Condition Room Temperature
Yield ≤47 μg
Show More Show Less
How should I prepare the MagMAX Plant RNA Kit (Cat. Nos. A33784, A33899, A47157) DNase I solution?

To prepare the MagMAX Plant RNA Kit (Cat. Nos. A33784, A33899) DNase I solution, add 440 µL of DNase I Reconstitution Buffer to each vial of DNase I (lyophilized) and incubate at room temperature for 5 minutes.

To prepare the MagMAX Plant RNA Kit (Cat. No. A47157) DNase I solution, add 10.5 mL of DNase I Reconstitution Buffer to the vial of DNase I (lyophilized) and incubate at room temperature for 5 minutes.

What kind of Polyvinylpyrrolidone (PVP) do you recommend using with MagMAX Plant DNA and RNA isolation kits since there are different kinds available?

For the MagMAX Plant DNA Isolation Kit, we recommend using PVP M.W. 40,000, Cat. No. J62417.A1 (https://www.thermofisher.com/order/catalog/product/J62417.A1).

For the MagMAX Plant RNA Isolation Kit, we recommend using Plant RNA Isolation Aid, Cat. No. AM9690 (https://www.thermofisher.com/order/catalog/product/AM9690).

Can I use the MagMAX Plant RNA Isolation Kit (Cat. No. A33784) to isolate small RNAs?

The MagMAX Plant RNA Isolation Kit has not been optimized for the isolation of small RNAs.

For Research Use Only. Not for use in diagnostic procedures.