missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™-96 Viral RNA Isolation Kit

Catalog No. AMB18365
Encompass
Change view
Click to view available options
No. of Reactions:
480
96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
AMB18365 480
AM1836 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AMB18365 Supplier Applied Biosystems™ Supplier No. AMB18365

This Applied Biosystems™ kit is for the isolation of viral RNA and DNA from diverse sample types (i.e., environmental samples and biofluids). Nucleic acids purified using this kit are ideal for PCR and RT-PCR applications.

Perfect for working with low viral concentrations, the kit contains sufficient reagents for 5 × 96 reactions.

Benefits of MagMAX™ Magnetic Bead-Based Purification:

Magnetic beads offer many benefits compared to other technologies for isolating RNA. Beads bind RNA more efficiently than glass fiber filters, resulting in higher and more consistent RNA yields. Additionally, because filters are not used, there is no risk of filter clogging due to cellular particulates in samples. Since only a small volume of magnetic beads is needed for high-efficiency binding, the bound RNA can be eluted in just 20–50μL of nuclease-free water, concentrating RNA from large, dilute samples.

Optimized for Isolation of Virus from Biofluids:

The Applied Biosystems™ MagMAX™ Viral RNA Isolation Kits have been optimized for use with biological fluids and cell-free samples such as serum, plasma, swabs, and cell culture media. These samples often present unique challenges when purifying RNA, including low viral titers, large volumes to be processed, and sample complexity. Without the need for organic solvents or RNA precipitation, the MagMAX™ Viral RNA Isolation Kit eliminates the problems often encountered with other RNA isolation methods such filter clogging, large elution volumes, and inconsistent yields. With this kit, DNA and RNA input levels ranging from 20 copies to 25 x 106 copies have been quantitatively recovered from plasma, serum, and milk. And even with less sample input, pathogen detection was as sensitive (equivalent CTs) or more sensitive (lower CTs) using nucleic acid isolated with the MagMAX™ kit when compared to nucleic acid isolated using another company's product. Downloadable protocols for the use of this kit with specific liquid handling systems are available at our Automation Resource.

Accessory Products:

Carrier RNA (SKU#4382878), Lysis/Binding Solution Concentrate (SKU#AM8500), Wash Solution 1 Concentrate (SKU#AM8504), and Wash Solution 2 Concentrate (SKU#AM8640) are components of the MagMAX™ Viral RNA Isolation Kits that may also be purchased separately. A 96-well magnetic ring stand is required to use this kit. The Magnetic Stand-96 (SKU#AM10027) and the 96-well Magnetic Ring Stand (SKU#AM10050) are designed for paramagnetic bead precipitation from standard 96- well, U-bottom microplates. The Magnetic-Ring Stand pellets the magnetic beads in a circle shape, enabling easy removal of supernatant by a robot or multichannel pipettors. For additional processing plates, Matrix 96-Well U Bottom Polypropylene Plates (Thermo Scientific, Item# 4917) are recommended. For an NVSL-validated version of this kit, please see the MagMAX™-96 AI/ND Viral RNA Isolation Kits (SKU#1929 and SKU#AM1835).

Specifications

Content And Storage Carrier RNA and Lysis Binding Enhancer is stored at –20°C; RNA Binding Beads are stored at 4°C. Wash Solution 2 Concentrate and Elution Buffer may be stored at either 4°C or room temperature. Lysis⁄Binding Solution Concentrate, Wash Solution 1 Concentrate, and Processing Plates and Lids are stored at room temperature. Nuclease-free water may be stored at any temperature.
Format 96-well Plate
Isolation Technology Magnetic Bead
Purification Time 45 min.
Sample Type Cell-free Biological Fluids, Liquid Samples (e.g. Serum), Viral Samples
Final Product Type Viral DNA⁄RNA
High-throughput Compatibility High-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
For Use With (Equipment) KingFisher™ Flex Magnetic Particle Processor, KingFisher 96
For Use With (Application) Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Northern Blotting
No. of Reactions 480
Product Line MagMAX
Quantity 5 x 96 Preps
Scale Maxi
Starting Material Amount Up to 50 μl
Target Viral DNA/RNA
Yield 5 ng
Show More Show Less
Are MagMax kits compatible with RNAlater reagent-treated samples?

Only MagMax96 for Microarrays Total RNA Isolation Kit (Cat. No. AM1839) is compatible, since it uses Trizol reagent and most of the salt carry-over from RNAlater reagent is eliminated during the lysis steps and partitioned from the aqueous RNA layer on top. For other MagMax kits, optimization is needed. High salt carry over is not optimal for MagMax RNA isolation steps.

Can I edit the manufactured KingFisher Flex protocols?

No, manufactured protocols are locked for editing, but they can be renamed using the ‘save as' function and edited as a new version.

Can I use magnetic beads from other vendors with the KingFisher Flex instrument?

For nucleic acid extraction, we recommend using our MagMAX kits and MagJET kits. Other magnetic beads should work well as long as they are 0.5 to 10 µm in size. However, you will have to write your own KingFisher protocols using the Thermo Scientific BindIt software and validate/optimize the protocol on your own.

I am working with liquid samples with high volume and want to extract nucleic acid from them using the KingFisher Flex instrument. Can I use more volume than the protocol advises?

Yes, this can be done by sample splitting. Set the protocol so that the magnet will collect samples from multiple wells before continuing to the next step. Be sure not to overload.

Can I scale up my reaction volume with the KingFisher Flex instrument?

Yes, reaction volumes can be scaled up, and all other volumes (i.e. wash volumes) will have to be increased by the same fold difference. It is generally not recommended to scale up reaction volumes more than 2-fold unless the plate's volume capacity will allow it.

Are all of the Applied Biosystems MagMAX kits validated for the KingFisher Flex Magnetic Particle Processor?

Yes, all MagMAX kits can be used with the KingFisher Flex Magnetic Particle Processor.


For Veterinary Use Only.