missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals MagFlex-96 aNAP System (RUO)
Description
- The MagFlex-96 aNAP System is capable of processing up to 96 samples per run, it delivers high-quality DNA or RNA from a wide range of sample types, including blood, tissues, cells, swabs, plants, microbes, and environmental samples
- With user-friendly preset programs and rapid processing, the system ensures consistent results and minimal hands-on time
Specifications
Specifications
| Product Type | Nucleic Acid Extraction System |
| Sample Type | Tissue, Blood, Bacteria, Fresh or Frozen Plant Tissue, Tissue Homogenate |
| Suitable Downstream Analysis | Next-Generation Sequencing |
| Volume (Metric) Processing | 50 to 1000 μL |
| Plates per Deck | 6 |
| Format | 96-well Plate |
| Capacity | 96 Samples/Run |