missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112175
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112175 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112175 Supplier Invitrogen™ Supplier No. PA5112175
Only null left

Rabbit Polyclonal Antibody

MAD2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP

Tight junction proteins (TJPs) belong to a family of membrane-associated guanylate kinase (MAGUK) homologs that are involved in the organization of epithelial and endothelial intercellular junctions. TJPs bind to the cytoplasmic C termini of junctional transmembrane proteins and link them to the actin cytoskeleton.

Specifications

Antigen MAD2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene MAD2L1
Gene Accession No. Q13257
Gene Alias AA673185; HsMAD2; J1256; MAD2; MAD2 (mitotic arrest deficient, homolog)-like 1; MAD2 (mitotic arrest deficient, yeast, homolog)-like 1; MAD2 mitotic arrest deficient-like 1; MAD2 mitotic arrest deficient-like 1 (yeast); Mad2a; MAD2L1; MAD2-like 1; MAD2-like protein 1; MadL1; Mitotic arrest deficient 2-like protein 1; mitotic arrest deficient, yeast, homolog-like 1; Mitotic MAD2 protein; Mitotic spindle assembly checkpoint protein MAD2A; Mitotic spindle checkpoint component MAD2; spindle checkpoint protein MAD2; YJL030W
Gene Symbols MAD2L1
Host Species Rabbit
Immunogen E. coli-derived Human MAD2L1 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4085
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less