missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAD2 Monoclonal Antibody (17D10)

Mouse Monoclonal Antibody
$826.48 - $1,249.99
Specifications
| Antigen | MAD2 |
|---|---|
| Clone | 17D10 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Description
Western blot detects a band of approximately 25 kDa (predicted molecular weight: 24.6 kDa).
Tight junction proteins (TJPs) belong to a family of membrane-associated guanylate kinase (MAGUK) homologs that are involved in the organization of epithelial and endothelial intercellular junctions. TJPs bind to the cytoplasmic C termini of junctional transmembrane proteins and link them to the actin cytoskeleton.Specifications
| MAD2 | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide | |
| Q13257 | |
| 4085 | |
| Recombinant human Mad2L1. | |
| Antibody |
| 17D10 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human, Mouse | |
| MAD2L1 | |
| AA673185; HsMAD2; J1256; MAD2; MAD2 (mitotic arrest deficient, homolog)-like 1; MAD2 (mitotic arrest deficient, yeast, homolog)-like 1; MAD2 mitotic arrest deficient-like 1; MAD2 mitotic arrest deficient-like 1 (yeast); Mad2a; MAD2L1; MAD2-like 1; MAD2-like protein 1; MadL1; Mitotic arrest deficient 2-like protein 1; mitotic arrest deficient, yeast, homolog-like 1; Mitotic MAD2 protein; Mitotic spindle assembly checkpoint protein MAD2A; Mitotic spindle checkpoint component MAD2; spindle checkpoint protein MAD2; YJL030W | |
| MAD2L1 | |
| Primary | |
| Protein G |