missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Machine/Equipment Label, Header: SAFETY FIRST, Legend: Carelessness is dangerous

Catalog No. 19112591
Change view
Click to view available options
Display Text:
Header: SAFETY FIRST, Legend: Carelessness is dangerous
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
19112591 Header: SAFETY FIRST, Legend: Carelessness is dangerous
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19112591 Supplier Brady™ Supplier No. 86905

Communicates important safety information and machine operational instructions. Brady Machine and Equipment Labels are made of durable, self-sticking polyester and most feature pictograms as well as text to inform safety procedures and potential hazards with machine operation.

  • Placed on machine or equipment to increase safety
  • Made of B-302 self sticking polyester
  • Available in size (W x H): 12.7 x 8.89cm (5 x 3.5 in.)

Specifications

Adhesive Type Acrylic, Permanent, Cold-temperature
Color Black, Green, White
Finish Gloss
Material Self-adhesive Polyester (B-302)
Product Type Machine/Equipment Label
Display Text Header: SAFETY FIRST, Legend: Carelessness is dangerous
Languages English
Quantity 5/Pk.
Shape Rectangle
Size (English) 5 x 3.5 in. (W x H)
Size (Metric) 127 x 88.9 mm (W x H)
Thickness (English) 0.01 in.
Show More Show Less