missing translation for 'onlineSavingsMsg'
Learn More

Cytiva MabSelect Xtra™ Chromatography Media

Catalog No. p-4673272
Change view
Click to view available options
Quantity:
25 mL
200 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002980 25 mL
45002979 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45002980 Supplier Cytiva Supplier No. 17526907
Only null left

Provides high purity capture and high dynamic binding capacity

  • High ligand selectivity and matrix hydrophilicity
  • Improved process economics through reduced raw materials costs and/or reduced number of cycles
  • Slightly lower pressure/flow properties compared to MabSelect™ SuRe™ and MabSelect

Specifications

Quantity 25 mL
For Use With (Application) For Develop to Meet The Demands of Increasing Levels of Expression in Monoclonal Antibody Feedstocks
Storage Requirements 2°C to 8°C
Binding Capacity 40 mg Human lgG/mL (at 2.4 min. residence time)
Product Type MabSelect Xtra Media
Flow Rate <300 cm/Hoμr
Particle Size 75 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Max. Pressure <200 kPa
Matrix Highly Cross-Linked Agarose
pH Range 2 to 12
Storage Buffer 20% Ethanol
Ligand Type Recombinant Protein A (E. coli)
Show More Show Less