missing translation for 'onlineSavingsMsg'
Learn More

Cytiva MabSelect Sure™ LX Chromatography Media

Catalog No. 45002568
Change view
Click to view available options
Quantity:
25 mL
200 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002568 25 mL
45002569 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45002568 Supplier Cytiva Supplier No. 17547401

Protein A affinity medium with high dynamic binding capacity which improves performance and cuts costs

Alkali tolerance allows for stability, thorough cleaning and reduction in contamination

  • Alkali-stabilized Protein A-derived (E.coli) Ligand
  • High dynamic binding capacity (DBC) for high titer cultures
  • Reduces risk of contamination and carryover
  • Alkali-tolerant rProtein A ligand withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH

Purification of monoclonal antibodies for clinical applications and process scale

Certifications

Certificate of Analysis

Specifications

Quantity 25 mL
Stability Stable in aqueous Buffers commonly used in Protein A Chromatography
For Use With (Application) For PurifiCatIon of Monoclonal Antibodies for Clinical AppliCatIons
Storage Requirements 2°C to 8°C
Binding Capacity 60 mg Human IgG/mL mediμm
Product Type MabSelect SuRe LX Media
Flow Rate 500 cm/Hoμr
Particle Size 85 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Includes Certificate of Analysis
Max. Pressure <2 bar
Matrix Highly Cross-Linked Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol
Ligand Type Alkali-stabilized Protein A-derived (E. coli)
Show More Show Less