missing translation for 'onlineSavingsMsg'
Learn More

Cytiva MabSelect™ Chromatography Media

Catalog No. 45002370
Change view
Click to view available options
Quantity:
25 mL
200 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002370 25 mL
45002978 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45002370 Supplier Cytiva Supplier No. 17519901

Protein A affinity medium designed for capturing MAbs from large sample volumes

  • High-flow agarose matrix allows for high flow velocities at production scale
  • BioProcess medium supported for industrial applications and well-established in approved processes
  • Low non-specific binding resulting in low levels of impurities in the product eluate pool
  • Oriented coupling of ligand and optimized matrix gives high dynamic binding capacity
  • Based on highly cross-linked agarose matrix with recombinant protein A ligand
  • Ligand is engineered to favor oriented coupling and expressed in E. coli and is free of components of mammalian origin
  • Hydrophilic nature of the base matrix ensures low levels of non-specific

  • Average particle size: 85μm (d50V)
  • pH Stability cleaning-in-place (CIP): 2 to 12
  • pH Stability working range: 3 to 10
  • Ligand: recombinant protein A (E.coli)
  • Matrix: highly cross-linked agarose
  • Binding capacity/mL chromatography medium: ∽30mg human lgG/mL at 2.4 min. residence time
  • Pressure/Flow specification: <500cm/hr., <200 kPa, BPG 300, bed height 20cm

Specifications

Quantity 25 mL
For Use With (Application) For Industrial AppliCatIons and Well-Established in Approved Processes
Storage Requirements 2°C to 8°C
Binding Capacity 30 mg Human lgG/mL (at 2.4 min. residence time)
Product Type MabSelect Media
Flow Rate <500 cm/Hoμr
Particle Size 85 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Includes Certificate of Analysis
Max. Pressure <200 kPa
Matrix Highly Cross-Linked Agarose
pH Range 3 to 10
Ligand Type Recombinant Protein A (E. coli)
Show More Show Less