missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ M1 OIML Class Advanced Weight Set (M1 PL C E)

Catalog No. 01912632
Change view
Click to view available options
Weight (Metric):
1 mg to 1 kg
1 mg to 2 kg
1 mg to 200 g
1 mg to 5 kg
1 to 50 g
1 to 500 g
1 to 500 mg
Includes:
12 Pieces; With certificate
23 Pieces; With certificate
25 Pieces; With certificate
27 Pieces; With certificate
28 Pieces; With certificate
8 Pieces; With certificate
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric) Includes
01912632 1 to 500 g 12 Pieces; With certificate
01912626 1 to 500 mg 12 Pieces; With certificate
01912627 1 mg to 200 g 23 Pieces; With certificate
01912628 1 mg to 1 kg 25 Pieces; With certificate
01912629 1 mg to 2 kg 27 Pieces; With certificate
01912630 1 mg to 5 kg 28 Pieces; With certificate
01912631 1 to 50 g 8 Pieces; With certificate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01912632 Supplier Mettler Toledo™ Supplier No. 30406451

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1 OIML quality weight set housed in a plastic case includes accessories for handling and cleaning, and a calibration certificate.

  • With calibration certificate
  • Traceability Certificate: Delivered with a Calibration Certificate in accordance with ISO/IEC 17025 requirements
  • Weight Calibration Performed by Robots: State-of-the-art robot technology minimizes errors and increases accuracy
  • Each knob weight has an adjusting cavity, which simplifies weight adjustment without the need for specialized equipment

Specifications

Product Type Test Weight
Material Stainless Steel
Case Material Plastic
Weight (Metric) 1 to 500 g
Density 7950 (± 140) kg/m3
Includes 12 Pieces; With certificate