missing translation for 'onlineSavingsMsg'
Learn More

m-Xylylenediamine 99.0+%, TCI America™

Catalog No. p-7126051
Change view
Click to view available options
Quantity:
100 g
25 g
Catalog No. Quantity
D012725G 25 g
D0127100G 100 g
2 options

Catalog No. D012725G

Supplier: TCI America D012725G

Only null left

Chemical Identifiers

CAS 1477-55-0
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00008119
InChI Key FDLQZKYLHJJBHD-UHFFFAOYSA-N
Synonym m-xylylenediamine, 1,3-benzenedimethanamine, 1,3-bis aminomethyl benzene, 1,3-xylylenediamine, 1,3-phenylenedimethanamine, m-xylylendiamin, 1,3-xylenediamine, mxda, m-phenylenebis methylamine, methylamine, m-phenylenebis
PubChem CID 15133
IUPAC Name [3-(aminomethyl)phenyl]methanamine
SMILES C1=CC(=CC(=C1)CN)CN

Specifications

Color Yellow
Boiling Point 248°C
Quantity 25 g
UN Number 2735
Formula Weight 136.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material m-Xylylenediamine
EINECSNumber (3)-0308
RTECSNumber PF8970000
TSCA Yes