missing translation for 'onlineSavingsMsg'
Learn More

m-Xylylene Diisocyanate 98.0+%, TCI America™

Catalog No. X002225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X002225G 25 g
X0022100G 100 g
X0022500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. X002225G Supplier TCI America Supplier No. X002225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3634-83-1
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD00019917
InChI Key RTTZISZSHSCFRH-UHFFFAOYSA-N
Synonym 1,3-Bis(isocyanatomethyl)benzene, Isocyanic Acid 1,3-Phenylenedimethylene Ester, m-Xylene Diisocyanate
PubChem CID 19262
IUPAC Name 1,3-bis(isocyanatomethyl)benzene
SMILES O=C=NCC1=CC(CN=C=O)=CC=C1

Specifications

Melting Point -7°C
Color Yellow
Boiling Point 126°C
Quantity 25 g
UN Number 2206
Formula Weight 188.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material m-Xylylene Diisocyanate
EINECSNumber (3)-2216
RTECSNumber NR0147000
TSCA Yes
Recommended Storage Refrigerator