missing translation for 'onlineSavingsMsg'
Learn More

m-Xylylene dibromide, 97%

Catalog No. AAB2495909
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2495909 10 g
AAB2495918 50 g
AAB2495930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2495909 Supplier Thermo Scientific Chemicals Supplier No. B2495909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 626-15-3 | C8H8Br2 | 263.96 g/mol

It is used in preparation of four novel dinuclear half-titanocenes with meta- and orthoxylene bridges as new metallocenes. It was used in the synthesis of dissymmetrical bis-macro cyclic salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in preparation of four novel dinuclear half-titanocenes with meta- and orthoxylene bridges as new metallocenes. It was used in the synthesis of dissymmetrical bis-macro cyclic salt.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents, active metals, bases.

Chemical Identifiers

CAS 626-15-3
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00000178
InChI Key OXHOPZLBSSTTBU-UHFFFAOYSA-N
Synonym 1,3-bis bromomethyl benzene, m-xylylene dibromide, alpha,alpha'-dibromo-m-xylene, benzene, 1,3-bis bromomethyl, m-bis bromomethyl benzene, m-xylylene bromide, ccris 1777, m-xylyene dibromide, m-xylene, .alpha.,.alpha.'-dibromo, m-.alpha.,.alpha.'-dibromoxylene
PubChem CID 69373
IUPAC Name 1,3-bis(bromomethyl)benzene
SMILES C1=CC(=CC(=C1)CBr)CBr

Specifications

Melting Point 74°C to 76°C
Quantity 10 g
UN Number UN3261
Beilstein 971085
Solubility Information Insoluble in water.
Formula Weight 263.96
Percent Purity 97%
Chemical Name or Material m-Xylylene dibromide
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 210-931-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only