missing translation for 'onlineSavingsMsg'
Learn More

M-Xylene, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023059
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023059 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023059 Supplier Merck Emd Millipore Supplier No. RTC00010120ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

Chemical Name or Material m-Xylene
Melting Point -48°C (literature)
CAS 108-38-3
Density 0.868 g/mL (at 25°C (literature))
Quantity 20 mL
Flash Point 25°C (Not Applicable)
Refractive Index n20/D 1.497 (literature)
Molecular Formula C8H10
Linear Formula C6H4(CH3)2
MDL Number MFCD00008536
Synonym m-Xylene; 1,3-Dimethylbenzene
Boiling Point 138°C to 139°C (literature)
Molecular Weight (g/mol) 106.17
Formula Weight 106.17
Grade Reference Material
Show More Show Less