missing translation for 'onlineSavingsMsg'
Learn More

m-Xylene, 99%

Catalog No. AAL03788AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL03788AP 500 mL
AAL03788AE 100 mL
AAL037880F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL03788AP Supplier Thermo Scientific Chemicals Supplier No. L03788AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 108-38-3 | C8H10 | 106.17 g/mol

m-Xylene is used in the production of isophthalic acid, which is a monomer and utilized in preparation of polyethylene terephthalate. It is used as an important starting material in the production of 2,4- and 2,6-xylidine. It is used as solvent in histology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Xylene is used in the production of isophthalic acid, which is a monomer and utilized in preparation of polyethylene terephthalate. It is used as an important starting material in the production of 2,4- and 2,6-xylidine. It is used as solvent in histology.

Solubility
Miscible with organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.  

Chemical Identifiers

CAS 108-38-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008536
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
Synonym m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150
PubChem CID 7929
ChEBI CHEBI:28488
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

Melting Point -47°C
Density 0.866
Quantity 500 mL
Flash Point 25°C (77°F)
Odor Mild
Refractive Index 1.497
UN Number UN1307
Beilstein 605441
Merck Index 14,10081
Boiling Point 138°C to 139°C
Solubility Information Miscible with organic solvents. Immiscible with water.
Formula Weight 106.17
Percent Purity 99%
Chemical Name or Material m-Xylene
Show More Show Less
Hazard Category H226-H312+H332-H315-H319
Hazard Statement GHS H Statement
H226-H312-H332-H315
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 203-576-3
RTECSNumber ZE2275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only