missing translation for 'onlineSavingsMsg'
Learn More

m-Xylene, 99+%, extra pure

Numéro de catalogue. p-3737748
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC180862500 250 mL Glass Bottle
AC180860010 1 L Glass Bottle
AC180860025 2.5 L Glass Bottle
AC180860100 10 L Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. AC180862500 Fournisseur Thermo Scientific Chemicals Code fournisseur 180862500
Il en reste null

CAS: 108-38-3 | C8H10 | 106.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 108-38-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008536
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
Synonym m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150
PubChem CID 7929
ChEBI CHEBI:28488
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Spécifications

Melting Point -48.0°C
Color Colorless
CAS Min % 99.0
CAS Max % 100.0
Density 0.8640g/mL
Quantity 250 mL
Flash Point 25°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4960 to 1.4980
Residue after Evaporation 0.005% max.
Specific Gravity 0.864
Assay Percent Range 99+%
Linear Formula C6H4(CH3)2
Viscosity 0.62 mPa.s (20°C)
Water 0.1% max.
Beilstein 05, 370
Merck Index 15, 10278
Boiling Point 139.0°C
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
Formula Weight 106.17
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material m-Xylene
Afficher plus de résultats Afficher moins de résultats
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only