missing translation for 'onlineSavingsMsg'
Learn More

m-Xylene, 98%, Spectrum™ Chemical

Catalog No. p-7166735
Change view
Click to view available options
Quantity:
20 L
200 L
4 L
500 mL
Packaging:
Amber Glass Bottle
Drum
Steel Can
Catalog No. Quantity Packaging
18611837 500 mL Amber Glass Bottle
18611836 4 L Amber Glass Bottle
18611834 20 L Steel Can
18611835 200 L Drum
4 options

Catalog No. 18611837

Supplier: Spectrum Chemical Mfg Cor X1015500MLGL

Only null left

Chemical Identifiers

CAS 108-38-3
Molecular Weight (g/mol) 106.17

Specifications

Boiling Point 137°C to 139°C
Quantity 500 mL
Packaging Amber Glass Bottle
Percent Purity 0.98
Grade Reagent
Chemical Name or Material m-Xylene

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.