missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Xylene-4-sulfonic Acid Hydrate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 61°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 186.23 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | m-Xylene-4-sulfonic Acid Hydrate |
Safety and Handling
| EINECSNumber | (9)-0317 |
| RTECSNumber | ZE5075000 |
| TSCA | Yes |