missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Tolyldiethanolamine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 70°C |
| Quantity | 500 g |
| Synonym | N,N-Bis(2-hydroxyethyl)-m-toluidine, 2,2′-(3-Methylphenylimino)diethanol, 2,2′-(m-Tolylimino)diethanol |
| Formula Weight | 195.26 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | m-Tolyldiethanolamine |
Safety and Handling
| EINECSNumber | (3)-0233 |
| RTECSNumber | KM3360000 |
| TSCA | Yes |