missing translation for 'onlineSavingsMsg'
Learn More

m-Tolyldiethanolamine 98.0+%, TCI America™

Catalog No. T031525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T031525G 25 g
T0315500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T031525G Supplier TCI America Supplier No. T031525G
Only null left

Chemical Identifiers

CAS 91-99-6
Molecular Formula C11H17NO2
Molecular Weight (g/mol) 195.26
MDL Number MFCD00020578
InChI Key VMNDRLYLEVCGAG-UHFFFAOYSA-N
PubChem CID 7073
IUPAC Name 2-[(2-hydroxyethyl)(3-methylphenyl)amino]ethan-1-ol
SMILES CC1=CC=CC(=C1)N(CCO)CCO

Specifications

Melting Point 70°C
Quantity 25 g
Synonym N,N-Bis(2-hydroxyethyl)-m-toluidine, 2,2′C-(3-Methylphenylimino)diethanol, 2,2′C-(m-Tolylimino)diethanol
Formula Weight 195.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material m-Tolyldiethanolamine
EINECSNumber (3)-0233
RTECSNumber KM3360000
TSCA Yes