missing translation for 'onlineSavingsMsg'
Learn More

m-Tolylacetylene, 97%

Catalog No. aal1718406
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
AAL1718406 5 g
AAL1718403 1 g
2 options

Catalog No. AAL1718406

Supplier: Thermo Scientific Chemicals L1718406

Only null left

CAS: 766-82-5 | C9H8 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 766-82-5
Molecular Formula C9H8
Molecular Weight (g/mol) 116.16
MDL Number MFCD01318174
InChI Key RENYIDZOAFFNHC-UHFFFAOYSA-N
PubChem CID 136600
IUPAC Name 1-ethynyl-3-methylbenzene
SMILES CC1=CC=CC(=C1)C#C

Specifications

Density 0.9
Boiling Point 170°C to 175°C
Flash Point 50°C (122°F)
Odor Pungent
Refractive Index 1.544
Quantity 5 g
UN Number UN3295
Beilstein 2038007
Synonym 3-ethynyltoluene, 3-methylphenylacetylene, m-tolylacetylene, 1-ethynyl-3-methyl-benzene, 3'-methylphenyl acetylene, benzene,1-ethynyl-3-methyl, benzene, 1-ethynyl-3-methyl, 3'-methylphenylacetylene, 3-methyl phenylacetylene, 3-tolylacetylene
Formula Weight 116.16
Percent Purity 97%
Chemical Name or Material m-Tolylacetylene
Show More Show Less
Hazard Category H226-H304-H315-H319-H335
Hazard Statement GHS H Statement
H304-H226-H319-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only