accessibility menu, dialog, popup

UserName

CH3HC

CAS RN 766-82-5

IUPAC Name: 1-ethynyl-3-methylbenzene
Synonyms: 1-ethynyl-3-methylbenzene
Molecular Weight (g/mol): 116.16
Molecular Formula: C9H8
InChi Key: RENYIDZOAFFNHC-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C#C
Boiling Point 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (4)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (5)