missing translation for 'onlineSavingsMsg'
Learn More

m-Tolyl Isothiocyanate 98.0+%, TCI America™

Catalog No. p-7131421
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04595G 5 g
I045925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04595G Supplier TCI America Supplier No. I04595G
Only null left

Chemical Identifiers

CAS 621-30-7
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.21
MDL Number MFCD00004807
InChI Key BDPQUWSFKCFOST-UHFFFAOYSA-N
Synonym m-tolyl isothiocyanate, 3-methylphenyl isothiocyanate, m-tolylisothiocyanate, benzene, 1-isothiocyanato-3-methyl, 3-tolyl isothiocyanate, isothiocyanic acid m-tolyl ester, 1-isothiocyanato-3-methyl-benzene, 1-isothiocyanato-3-methylbenzol, 3-isothiocyanatotoluene, acmc-209mzt
PubChem CID 69304
IUPAC Name 1-isothiocyanato-3-methylbenzene
SMILES CC1=CC=CC(=C1)N=C=S

Specifications

Color Yellow
Boiling Point 240°C
Quantity 5 g
UN Number 2810
Formula Weight 149.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material m-Tolyl Isothiocyanate