missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Toluoyl Chloride 98.0+%, TCI America™
$140.54 - $363.85
Chemical Identifiers
| CAS | 1711-06-4 |
|---|---|
| Molecular Formula | C8H7ClO |
| Molecular Weight (g/mol) | 154.593 |
| MDL Number | MFCD00000681 |
| InChI Key | YHOYYHYBFSYOSQ-UHFFFAOYSA-N |
| Synonym | m-toluoyl chloride, m-methylbenzoyl chloride, 3-toluoyl chloride, toluoyl chloride, benzoyl chloride, 3-methyl, m-toluyl chloride, meta methyl benzoyl chloride, 3-methylbenzoylchloride, 3-toluoylchloride, unii-w2vjw4350k |
| PubChem CID | 74375 |
| IUPAC Name | 3-methylbenzoyl chloride |
| SMILES | CC1=CC=CC(=C1)C(=O)Cl |
Chemical Identifiers
| 1711-06-4 | |
| 154.593 | |
| YHOYYHYBFSYOSQ-UHFFFAOYSA-N | |
| 74375 | |
| CC1=CC=CC(=C1)C(=O)Cl |
| C8H7ClO | |
| MFCD00000681 | |
| m-toluoyl chloride, m-methylbenzoyl chloride, 3-toluoyl chloride, toluoyl chloride, benzoyl chloride, 3-methyl, m-toluyl chloride, meta methyl benzoyl chloride, 3-methylbenzoylchloride, 3-toluoylchloride, unii-w2vjw4350k | |
| 3-methylbenzoyl chloride |
Specifications
| 1711-06-4 | |
| Yellow | |
| C8H7ClO | |
| 25 g | |
| m-toluoyl chloride, m-methylbenzoyl chloride, 3-toluoyl chloride, toluoyl chloride, benzoyl chloride, 3-methyl, m-toluyl chloride, meta methyl benzoyl chloride, 3-methylbenzoylchloride, 3-toluoylchloride, unii-w2vjw4350k | |
| CC1=CC=CC(=C1)C(=O)Cl | |
| 154.593 | |
| 154.59 | |
| Liquid |
| -23°C | |
| 220°C | |
| MFCD00000681 | |
| 3265 | |
| YHOYYHYBFSYOSQ-UHFFFAOYSA-N | |
| 3-methylbenzoyl chloride | |
| 74375 | |
| ≥98.0% (GC,T) | |
| m-Toluoyl Chloride |
Safety and Handling
EINECSNumber : (3)-3481
RTECSNumber : DM6644000
TSCA : Yes