accessibility menu, dialog, popup

UserName

CH3ClO

CAS RN 1711-06-4

IUPAC Name: 3-methylbenzoyl chloride
Synonyms: benzoyl chloride, 3-methyl-
Molecular Weight (g/mol): 154.59
Molecular Formula: C8H7ClO
InChi Key: YHOYYHYBFSYOSQ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C(Cl)=O
Boiling Point 
  • (5)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (4)