missing translation for 'onlineSavingsMsg'
Learn More

m-Toluoyl chloride, 99%

CAS: 1711-06-4 | C8H7ClO | 154.6 g/mol

$164.96 - $164.96

Chemical Identifiers

CAS 1711-06-4
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.6
MDL Number MFCD00000681
InChI Key YHOYYHYBFSYOSQ-UHFFFAOYSA-N
Synonym m-toluoyl chloride, m-methylbenzoyl chloride, 3-toluoyl chloride, toluoyl chloride, benzoyl chloride, 3-methyl, m-toluyl chloride, meta methyl benzoyl chloride, 3-methylbenzoylchloride, 3-toluoylchloride, unii-w2vjw4350k
PubChem CID 74375
IUPAC Name 3-methylbenzoyl chloride
SMILES CC1=CC=CC(=C1)C(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC153411000
SDS View Documents
Thermo Scientific Chemicals
153411000
100 g Glass bottle
Each for $164.96
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1711-06-4
154.6
YHOYYHYBFSYOSQ-UHFFFAOYSA-N
74375
CC1=CC=CC(=C1)C(=O)Cl
C8H7ClO
MFCD00000681
m-toluoyl chloride, m-methylbenzoyl chloride, 3-toluoyl chloride, toluoyl chloride, benzoyl chloride, 3-methyl, m-toluyl chloride, meta methyl benzoyl chloride, 3-methylbenzoylchloride, 3-toluoylchloride, unii-w2vjw4350k
3-methylbenzoyl chloride

Specifications

1711-06-4
100.0
1.1700g/mL
76°C
99%
C8H7ClO
CH3C6H4COCl
100 g
1.17
Solubility in water: decomposes.
CC1=CC=CC(=C1)C(=O)Cl
154.6
154.6
Liquid
98.5
Colorless to Brown
86.0°C (5.0 mmHg)
Authentic
Glass bottle
1.5475 to 1.5495
MFCD00000681
09, 477
m-toluoyl chloride, m-methylbenzoyl chloride, 3-toluoyl chloride, toluoyl chloride, benzoyl chloride, 3-methyl, m-toluyl chloride, meta methyl benzoyl chloride, 3-methylbenzoylchloride, 3-toluoylchloride, unii-w2vjw4350k
YHOYYHYBFSYOSQ-UHFFFAOYSA-N
3-methylbenzoyl chloride
74375
99%
m-Toluoyl chloride, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

EINECSNumber : 216-976-8

RTECSNumber : DM6644000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only