missing translation for 'onlineSavingsMsg'
Learn More

m-Tolunitrile, 99%

CAS: 620-22-4 | C8H7N | 117.15 g/mol

$61.70 - $686.87

Chemical Identifiers

CAS 620-22-4
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.15
MDL Number MFCD00001808
InChI Key BOHCMQZJWOGWTA-UHFFFAOYSA-N
Synonym m-tolunitrile, 3-cyanotoluene, benzonitrile, 3-methyl, m-cyanotoluene, 3-tolunitrile, m-toluonitrile, m-tolylnitrile, m-toluenenitrile, m-methylbenzonitrile, m-tolynitrile
PubChem CID 12104
IUPAC Name 3-methylbenzonitrile
SMILES CC1=CC=CC(=C1)C#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC158010050
SDS View Documents
Thermo Scientific Chemicals
158010050
5 g Glass bottle
Each for $61.70
Only null left
 
AC158011000
SDS View Documents
Thermo Scientific Chemicals
158011000
100 g Glass bottle
Each for $133.05
Only null left
 
AC158015000
SDS View Documents
Thermo Scientific Chemicals
158015000
500 g Glass bottle
Each for $686.87
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

620-22-4
117.15
BOHCMQZJWOGWTA-UHFFFAOYSA-N
12104
CC1=CC=CC(=C1)C#N
C8H7N
MFCD00001808
m-tolunitrile, 3-cyanotoluene, benzonitrile, 3-methyl, m-cyanotoluene, 3-tolunitrile, m-toluonitrile, m-tolylnitrile, m-toluenenitrile, m-methylbenzonitrile, m-tolynitrile
3-methylbenzonitrile

Specifications

620-22-4
100.0
Colorless to Yellow
99°C to 101°C (20.0 mmHg)
Authentic
Glass bottle
1.5246 to 1.5266
MFCD00001808
09, 477
m-tolunitrile, 3-cyanotoluene, benzonitrile, 3-methyl, m-cyanotoluene, 3-tolunitrile, m-toluonitrile, m-tolylnitrile, m-toluenenitrile, m-methylbenzonitrile, m-tolynitrile
BOHCMQZJWOGWTA-UHFFFAOYSA-N
3-methylbenzonitrile
12104
99%
m-Tolunitrile
98.5
-23°C
0.9700g/mL
86°C
99%
C8H7N
CH3C6H4CN
5 g
0.97
Solubility in water: .. Other solubilities: solunle in methanol,ethanol,dioxane
CC1=CC=CC(=C1)C#N
117.15
117.15
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective cloth

GHS Signal Word: Warning

EINECSNumber : 210-631-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only