missing translation for 'onlineSavingsMsg'
Learn More

m-Tolunitrile, 99%

CAS: 620-22-4 | C8H7N | 117.15 g/mol

Supplier:  Thermo Scientific Chemicals 158011000

 View more versions of this product

Catalog No. AC158011000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

m-Tolunitrile
98.5
-23°C
0.9700g/mL
86°C
99%
C8H7N
CH3C6H4CN
100 g
0.97
Solubility in water: .. Other solubilities: solunle in methanol,ethanol,dioxane
CC1=CC=CC(=C1)C#N
117.15
117.15
Liquid
620-22-4
100.0
Colorless to Yellow
99°C to 101°C (20.0 mmHg)
Authentic
Glass bottle
1.5246 to 1.5266
MFCD00001808
09, 477
m-tolunitrile, 3-cyanotoluene, benzonitrile, 3-methyl, m-cyanotoluene, 3-tolunitrile, m-toluonitrile, m-tolylnitrile, m-toluenenitrile, m-methylbenzonitrile, m-tolynitrile
BOHCMQZJWOGWTA-UHFFFAOYSA-N
3-methylbenzonitrile
12104
99%

Chemical Identifiers

620-22-4
117.15
BOHCMQZJWOGWTA-UHFFFAOYSA-N
12104
CC1=CC=CC(=C1)C#N
C8H7N
MFCD00001808
m-tolunitrile, 3-cyanotoluene, benzonitrile, 3-methyl, m-cyanotoluene, 3-tolunitrile, m-toluonitrile, m-tolylnitrile, m-toluenenitrile, m-methylbenzonitrile, m-tolynitrile
3-methylbenzonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective cloth

GHS Signal Word: Warning

EINECSNumber : 210-631-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only