missing translation for 'onlineSavingsMsg'
Learn More

m-Toluidine 98.0+%, TCI America™

Catalog No. T0298500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0298500ML 500 mL
T029825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0298500ML Supplier TCI America Supplier No. T0298500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-44-1
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00007808
InChI Key JJYPMNFTHPTTDI-UHFFFAOYSA-N
Synonym m-toluidine, 3-toluidine, benzenamine, 3-methyl, 3-aminotoluene, m-methylaniline, m-tolylamine, m-toluidin, m-aminotoluene, m-methylbenzenamine, 3-methylbenzenamine
PubChem CID 7934
IUPAC Name 3-methylaniline
SMILES CC1=CC=CC(N)=C1

Specifications

Color Yellow
Boiling Point 203°C
Quantity 500 mL
UN Number 1708
Formula Weight 107.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material m-Toluidine
EINECSNumber (3)-0186
RTECSNumber XU2800000
TSCA Yes
Recommended Storage Refrigerator