missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Toluidine, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | -30.0°C |
| Color | White |
| Density | 0.9800g/mL |
| Boiling Point | 203.0°C to 204.0°C |
| Flash Point | 86°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass Bottle |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause damage to organs through prolonged or repeated exposure. Toxic if inhaled. Toxic in contact with skin. Very toxic to aquatic life. Toxic if swallowed. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fre |
| EINECSNumber | 203-583-1 |
| RTECSNumber | XU2800000 |
| TSCA | TSCA |
RUO – Research Use Only