missing translation for 'onlineSavingsMsg'
Learn More

m-Thiocresol, 97%

Catalog No. AAB2381814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2381814 25 g
AAB2381806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2381814 Supplier Thermo Scientific Chemicals Supplier No. B2381814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 108-40-7 | C7H8S | 124.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-40-7
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.20
MDL Number MFCD00004843
InChI Key WRXOZRLZDJAYDR-UHFFFAOYSA-N
Synonym m-thiocresol, m-toluenethiol, 3-methylthiophenol, 3-thiocresol, benzenethiol, 3-methyl, m-methylbenzenethiol, m-methylthiophenol, 3-mercaptotoluene, m-tolylmercaptan, m-mercaptotoluene
PubChem CID 7930
IUPAC Name 3-methylbenzenethiol
SMILES CC1=CC=CC(S)=C1

Specifications

Melting Point <-20°C
Density 1.044
Boiling Point 196°C
Flash Point 72°C (161°F)
Refractive Index 1.5762
Quantity 25 g
UN Number UN2810
Beilstein 1851959
Sensitivity Air Sensitive
Merck Index 14,9325
Formula Weight 124.21
Percent Purity 97%
Chemical Name or Material m-Thiocresol
Show More Show Less
Hazard Category H227-H301+H311+H331
Hazard Statement GHS H Statement
H331-H318-H227-H302-H312-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 203-578-4
RTECSNumber XT8700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only