missing translation for 'onlineSavingsMsg'
Learn More

m-Nitroacetophenone, 98%

Catalog No. p-172613
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC128331000 100 g Plastic bottle
AC128335000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC128331000 Supplier Thermo Scientific Chemicals Supplier No. 128331000
Only null left

CAS: 121-89-1 | C8H7NO3 | 165.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121-89-1
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00007259
InChI Key ARKIFHPFTHVKDT-UHFFFAOYSA-N
Synonym 3'-nitroacetophenone, 3-nitroacetophenone, m-nitroacetophenone, 1-3-nitrophenyl ethanone, ethanone, 1-3-nitrophenyl, acetophenone, 3'-nitro, m-acetylnitrobenzene, usaf ma-1, methyl 3-nitrophenyl ketone, 3-nitroacetofenon
PubChem CID 8494
IUPAC Name 1-(3-nitrophenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 76.0°C to 79.0°C
Boiling Point 201.0°C to 203.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula O2NC6H4COCH3
Quantity 100 g
Beilstein 07, 288
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 165.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material m-Nitroacetophenone, 98%
Show More Show Less
EINECSNumber 204-504-3
RTECSNumber AM9625000
TSCA TSCA

RUO – Research Use Only