accessibility menu, dialog, popup

UserName

CH3ONOO

CAS RN 121-89-1

IUPAC Name: 1-(3-nitrophenyl)ethan-1-one
Synonyms: 3-nitroacetophenone
Molecular Weight (g/mol): 165.15
Molecular Formula: C8H7NO3
InChi Key: ARKIFHPFTHVKDT-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC(=C1)[N+]([O-])=O
Boiling Point 
  • (2)
  • (5)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)