missing translation for 'onlineSavingsMsg'
Learn More

m,m'-Ditolylamine 98.0+%, TCI America™

Catalog No. D15125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15125G 5 g
D151225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15125G Supplier TCI America Supplier No. D15125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 626-13-1
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.281
MDL Number MFCD00059315
InChI Key CWVPIIWMONJVGG-UHFFFAOYSA-N
Synonym di-m-tolylamine, 3,3'-dimethyldiphenylamine, m,m'-ditolylamine, dim-tolylamine, bis 3-methylphenyl amine, 3-methyl-n-3-methylphenyl aniline, benzenamine, 3-methyl-n-3-methylphenyl, di-m-tolyl-amine, 3,3'-ditolylamine, pubchem8963
PubChem CID 7016139
IUPAC Name 3-methyl-N-(3-methylphenyl)aniline
SMILES CC1=CC(=CC=C1)NC2=CC=CC(=C2)C

Specifications

Melting Point -12°C
Boiling Point 200°C
Quantity 5 g
Formula Weight 197.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material m,m′-Ditolylamine