missing translation for 'onlineSavingsMsg'
Learn More

m-Cresol 98.0+%, TCI America™

Catalog No. C0401500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0401500G 500 g
C040125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0401500G Supplier TCI America Supplier No. C0401500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-39-4
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002302
InChI Key RLSSMJSEOOYNOY-UHFFFAOYSA-N
Synonym m-cresol, metacresol, meta-cresol, phenol, 3-methyl, 3-hydroxytoluene, m-methylphenol, 3-cresol, m-kresol, m-oxytoluene, m-toluol
PubChem CID 342
ChEBI CHEBI:17231
IUPAC Name 3-methylphenol
SMILES CC1=CC(=CC=C1)O

Specifications

Color Yellow
Boiling Point 202°C
Quantity 500 g
UN Number 2076
Formula Weight 108.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material m-Cresol
EINECSNumber (3)-0499&(4)-0057
RTECSNumber GO6125000
TSCA Yes