missing translation for 'onlineSavingsMsg'
Learn More

m-Cresol Purple Indicator, 0.04% (w/v), Ricca Chemical

$155.00 - $155.00

Chemical Identifiers

CAS 62625-31-4
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
2400-16
View Documents
Ricca Chemical Company
240016
500 mL Natural Poly Bottle
Each for $155.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

62625-31-4

Specifications

62625-31-4,7732-18-5
100.0,0.04
0.04% W/V
Natural Poly Bottle
Red/Orange
500 mL
m-Cresol Purple Indicator
98.0,0.04
0.04% (w/v)
Indicator
Aqueous Solution
7 to 9
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'