missing translation for 'onlineSavingsMsg'
Learn More

m-Cresol, High Purity, 99%, Spectrum™ Chemical

C1367, 108-39-4, C7H8O

Supplier:  Spectrum Chemical Mfg Cor C13671KGBL

 View more versions of this product

Catalog No. 18604240


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical m-Cresol, High Purity is a colorless organic compound and liquid. It is derived from phenol and is commonly extracted from coal tar. It is also known as 3-methylphenol. High purity products supplied by Spectrum are indicative of a grade suitable for research purposes and typically are not suitable for human consumption or therapeutic use.
Specifications

Specifications

108-39-4
C7H8O
1 kg
CC1=CC=CC(O)=C1
108.14
High Purity
100%
UN2076
RLSSMJSEOOYNOY-UHFFFAOYSA-N
3-methylphenol
99%
Amber Glass Bottle

Chemical Identifiers

108-39-4
108.14
3-methylphenol
C7H8O
RLSSMJSEOOYNOY-UHFFFAOYSA-N
CC1=CC=CC(O)=C1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions