missing translation for 'onlineSavingsMsg'
Learn More

m-Cresol, High Purity, 99%, Spectrum™ Chemical

Catalog No. p-7157157
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Catalog No. Quantity Packaging
18604238 100 g Amber Glass Bottle
18604239 500 g Amber Glass Bottle
18604241 20 L Poly Drum
18604242 200 L Drum
18604240 1 kg Amber Glass Bottle
5 options

Catalog No. 18604238

Supplier: Spectrum Chemical Mfg Cor C1367100GM

Only null left

C1367, 108-39-4, C7H8O

Spectrum™ Chemical m-Cresol, High Purity is a colorless organic compound and liquid. It is derived from phenol and is commonly extracted from coal tar. It is also known as 3-methylphenol. High purity products supplied by Spectrum are indicative of a grade suitable for research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 108-39-4
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
InChI Key RLSSMJSEOOYNOY-UHFFFAOYSA-N
IUPAC Name 3-methylphenol
SMILES CC1=CC=CC(O)=C1

Specifications

CAS Max % 100%
UN Number UN2076
Quantity 100 g
Percent Purity 99%
Grade High Purity
Packaging Amber Glass Bottle