missing translation for 'onlineSavingsMsg'
Learn More

m-Cresol, MilliporeSigma™

Catalog No. M8096919030
Change view
Click to view available options
Quantity:
30 kg
Packaging:
Steel Drum w/PE Liner
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M8096919030 30 kg Steel Drum w/PE Liner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M8096919030 Supplier Technidata Supplier No. 8.09691.9030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-39-4
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
InChI Key RLSSMJSEOOYNOY-UHFFFAOYSA-N
Synonym m-cresol, metacresol, meta-cresol, phenol, 3-methyl, 3-hydroxytoluene, m-methylphenol, 3-cresol, m-kresol, m-oxytoluene, m-toluol
PubChem CID 342
ChEBI CHEBI:17231
IUPAC Name 3-methylphenol
SMILES CC1=CC(=CC=C1)O

Specifications

Melting Point 8°C to 12°C
Density 1.03g/cm3
Boiling Point 202.6°C
Ignition Point 557.85°C (1036.1°F)
Odor Sweet
Assay Percent Range ≥90%
Packaging Steel Drum w/PE Liner
Linear Formula C7H8O
Quantity 30 kg
UN Number 2076
Solubility Information Partially soluble in water
Formula Weight 108.15
Physical Form Liquid
Chemical Name or Material m-Cresol
Show More Show Less
DOTInformation DOT Class: 6.1 (8), Packaging Group: II
Recommended Storage Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.