missing translation for 'onlineSavingsMsg'
Learn More

m-Cresol, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187432
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020671 1 mL
111019527 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020671 Supplier Merck Emd Millipore Supplier No. 659961MLF
Only null left

m-Cresol is an isomer form of Cresol and is mainly used as an intermediate in the manufacture of chemicals. It is toxic in nature. It is a component of coal tar creosote which is used for wood preservation. It also possess bactericide and fungicide property.

Featured Industries: Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 108-39-4
Ignition Point 1036°F
Vapor Pressure <1 mmHg (20°C)
Vapor Density 3.72 (vs. air)
Density 1.034 g/mL (at 25°C)
Flash Point 86°C
Physical Form Neat
Linear Formula CH3C6H4 OH
MDL Number MFCD00002302
Melting Point 8°C to 10°C
Boiling Point 203°C
Synonym 1-Hydroxy-3-methylbenzene; 3-Hydroxytoluene; 3-Methylphenol
Type Analytical Standard
Quantity 1 mL
Formula Weight 108.14
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less