missing translation for 'onlineSavingsMsg'
Learn More
Learn More
M-cadherin Monoclonal Antibody (K92056_12G8), Invitrogen™
Mouse Monoclonal Antibody
$1421.98 - $2134.69
Specifications
| Antigen | M-cadherin |
|---|---|
| Clone | K92056_12G8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: VLEGAVPGTY VTRAEATDAD DPETDNAALR FSILQQGSPE LFSIDELTGE IRTVQVGLDR EVVAVYNLTL QVADMSGDGL TATASAIITL DDINDNAPEF TRDEFFMEAI EAVSGVDVGR LEVEDRDLPG SPNWVARFTI LEGDPDGQFT IRTDPKTNEG VLSIVKALDY ESCEHYELKV SVQNEAPLQA AALRAERGQA KVRVHVQDTN
M-cadherin Monoclonal antibody specifically detects M-cadherin in Human, Mouse samples. It is validated for Western BlotSpecifications
| M-cadherin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AI323380; cadherin 15; cadherin 15, type 1, M-cadherin (myotubule); cadherin-14; Cadherin-15; cadherin-3; Cdh14; Cdh15; CDH3; CDHM; CRSBP-1; HAR; LYVE-1; M cadherin; Mcad; M-cadherin; MRD3; Muscle cadherin; muscle-cadherin; XLKD1 | |
| Recombinant human CDH15 (immunogen range is 161-370). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| CDH15 |
| K92056_12G8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| P33146, P55291 | |
| 1013, 12555 | |
| IgG2b | |
| Protein A | |
| Antibody |