missing translation for 'onlineSavingsMsg'
Learn More

m-Anisidine Hydrochloride 99.0+%, TCI America™

$77.17 - $234.87

Chemical Identifiers

CAS 27191-09-9
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.61
MDL Number MFCD00060220
InChI Key FNCKZWCTEPKMQV-UHFFFAOYSA-N
Synonym 3-Methoxyaniline Hydrochloride
PubChem CID 12232010
IUPAC Name 3-methoxyaniline hydrochloride
SMILES Cl.COC1=CC=CC(N)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A04885G
View Documents
TCI America
A04885G
5 g N/A
Only null left
 
A048825G
View Documents
TCI America
A048825G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

27191-09-9
159.61
FNCKZWCTEPKMQV-UHFFFAOYSA-N
12232010
Cl.COC1=CC=CC(N)=C1
C7H10ClNO
MFCD00060220
3-Methoxyaniline Hydrochloride
3-methoxyaniline hydrochloride

Specifications

27191-09-9
C7H10ClNO
5 g
3-Methoxyaniline Hydrochloride
Cl.COC1=CC=CC(N)=C1
159.61
159.61
Crystalline Powder
174°C
MFCD00060220
2811
FNCKZWCTEPKMQV-UHFFFAOYSA-N
3-methoxyaniline hydrochloride
12232010
≥99.0% (HPLC,N)
m-Anisidine Hydrochloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0682

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'