missing translation for 'onlineSavingsMsg'
Learn More

m-Anisaldehyde, 97%

Catalog No. p-4510323
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156760250 25 mL Glass bottle
AC156761000 100 mL Glass bottle
AC156765000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC156760250 Supplier Thermo Scientific Chemicals Supplier No. 156760250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 591-31-1 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 591-31-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
InChI Key WMPDAIZRQDCGFH-UHFFFAOYSA-N
Synonym m-anisaldehyde, 3-anisaldehyde, benzaldehyde, 3-methoxy, m-methoxybenzaldehyde, metamethoxybenzaldehyde, 3-methoxy-benzaldehyde, unii-8zao7s0ivh, ccris 960, meta-anisaldehyde, 8zao7s0ivh
PubChem CID 11569
IUPAC Name 3-methoxybenzaldehyde
SMILES COC1=CC=CC(=C1)C=O

Specifications

Color Yellow
Density 1.1190g/mL
Boiling Point 143.0°C (50.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5500 to 1.5540
Linear Formula CH3OC6H4CHO
Quantity 25 mL
Beilstein 08,59
Specific Gravity 1.119
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether
Formula Weight 136.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material m-Anisaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 209-712-8

RUO – Research Use Only