missing translation for 'onlineSavingsMsg'
Learn More

m-Acetotoluidide, Thermo Scientific™

$58.92 - $58.92

Chemical Identifiers

CAS 537-92-8
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00014962
InChI Key ALMHSXDYCFOZQD-UHFFFAOYSA-N
Synonym 3'-methylacetanilide, n-acetyl-m-toluidine, m-acetotoluidide, 3-methylacetanilide, n-3-methylphenyl acetamide, m-acetotoluide, 3-acetamidotoluene, n-m-tolylacetamide, m-methylacetanilide, aceto-m-aminotoluene
PubChem CID 10843
IUPAC Name N-(3-methylphenyl)acetamide
SMILES CC1=CC(=CC=C1)NC(=O)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC391300250
SDS View Documents
Thermo Scientific Chemicals
391300250
25 g Glass bottle
Each for $58.92
Only null left
 
Description

Description

Specifications

Chemical Identifiers

537-92-8
149.19
ALMHSXDYCFOZQD-UHFFFAOYSA-N
10843
CC1=CC(=CC=C1)NC(=O)C
C9H11NO
MFCD00014962
3'-methylacetanilide, n-acetyl-m-toluidine, m-acetotoluidide, 3-methylacetanilide, n-3-methylphenyl acetamide, m-acetotoluide, 3-acetamidotoluene, n-m-tolylacetamide, m-methylacetanilide, aceto-m-aminotoluene
N-(3-methylphenyl)acetamide

Specifications

537-92-8
303°C
C9H11NO
MFCD00014962
15, 72
ALMHSXDYCFOZQD-UHFFFAOYSA-N
N-(3-methylphenyl)acetamide
10843
m-Acetotoluidide
65°C to 67°C
Glass bottle
CH3C6H4NHC(O)CH3
25 g
3'-methylacetanilide, n-acetyl-m-toluidine, m-acetotoluidide, 3-methylacetanilide, n-3-methylphenyl acetamide, m-acetotoluide, 3-acetamidotoluene, n-m-tolylacetamide, m-methylacetanilide, aceto-m-aminotoluene
CC1=CC(=CC=C1)NC(=O)C
149.19
149.19
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-678-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only