missing translation for 'onlineSavingsMsg'
Learn More

m-Acetotoluidide, Thermo Scientific™

Catalog No. AC391300250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC391300250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC391300250 Supplier Thermo Scientific Chemicals Supplier No. 391300250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 537-92-8
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00014962
InChI Key ALMHSXDYCFOZQD-UHFFFAOYSA-N
Synonym 3'-methylacetanilide, n-acetyl-m-toluidine, m-acetotoluidide, 3-methylacetanilide, n-3-methylphenyl acetamide, m-acetotoluide, 3-acetamidotoluene, n-m-tolylacetamide, m-methylacetanilide, aceto-m-aminotoluene
PubChem CID 10843
IUPAC Name N-(3-methylphenyl)acetamide
SMILES CC1=CC(=CC=C1)NC(=O)C

Specifications

Melting Point 65°C to 67°C
Boiling Point 303°C
Packaging Glass bottle
Linear Formula CH3C6H4NHC(O)CH3
Quantity 25 g
Merck Index 15, 72
Formula Weight 149.19
Chemical Name or Material m-Acetotoluidide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-678-1

RUO – Research Use Only