missing translation for 'onlineSavingsMsg'
Learn More

m-(2-Fluorophenoxy)toluene, TCI America™

Catalog No. F03755G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F03755G Supplier TCI America Supplier No. F03755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78850-78-9
Molecular Formula C13H11FO
Molecular Weight (g/mol) 202.228
MDL Number MFCD01321169
InChI Key LTTPKDJFHJHNPA-UHFFFAOYSA-N
Synonym 2-Fluoro-3′C-methyldiphenyl Ether, 2-Fluorophenyl m-Tolyl Ether
PubChem CID 20570698
IUPAC Name 1-fluoro-2-(3-methylphenoxy)benzene
SMILES CC1=CC(=CC=C1)OC2=CC=CC=C2F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 202.23
Physical Form Liquid
Chemical Name or Material m-(2-Fluorophenoxy)toluene