missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Lyph-Seal Tray Dryer

The Lyph-Seal Tray Dryer can freeze dry up to 8 bags and seal under vacuum, allowing for long term storage or secure shipping.

Specifications

No. of Shelves 4
Temperature Range (Metric) Up to 60°C
View More Specs

Products 3
Catalog Number Mfr. No. Plug Type Voltage Price Quantity  
Catalog Number Mfr. No. Plug Type Voltage Price Quantity  
16-985-647
View Documents
Labconco™
780401010
North America 230 V N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
16-985-648
View Documents
Labconco™
780401000
North America 115 V N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
16-985-641
View Documents
Labconco™
780401090
All except North America 230 V N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Includes 4 heated shelves with control from ambient to 50°C for optimized lyophilization.
Specifications

Specifications

4
Up to 60°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'