Learn More
Invitrogen™ Lyo-ready Enzymes and Reagents for Recombinase Polymerase Amplification (RPA)
Invitrogen Lyo-ready RPA reagents are designed for fast, sensitive, and specific DNA amplification through isothermal recombinase polymerase amplification (RPA) reactions. Lyo-ready RPA reagents are available in stand-alone and kit formats.
Supplier: Invitrogen™ A72124
Description
Invitrogen Lyo-ready recombinase polymerase amplification (RPA) reagents are designed for fast, sensitive, and specific DNA amplification through isothermal RPA reactions. Lyo-ready RPA reagents are available in stand-alone and kit formats. Reverse transcriptase can be added to the RPA reaction for amplification of RNA targets. All reagents are glycerol-free, which enables assay lyophilization.
Features of Lyo-ready RPA products
- Enhanced sensitivity down to 1 copy using optimized (RT-)RPA reaction buffer
- Robust and specific RNA/DNA amplification
- Amplification from inhibitor-containing samples
- Fast target amplification in as little as 20 minutes
- Ability to easily optimize RPA or RT-RPA reactions
Kit components
The Lyo-ready RPA Kit includes all required components for an RPA reaction:
- Lyo-ready T4 UvsX Protein—produced from E. coli cells bearing cloned uvsX gene derived from bacteriophage T4
- Lyo-ready T4 UvsY Protein—a recombination mediator protein that stimulates homologous recombination performed by the UvsX recombinase
- Lyo-ready Bst DNA Polymerase (Cat. No. A56655)—a proprietary mutant of Bst DNA Polymerase, Large Fragment, with fast reaction speed, increased sensitivity, and tolerance to inhibitors
- Lyo-ready T4 Gene 32 Protein—a single-stranded DNA binding protein produced from E. coli cells carrying cloned bacteriophage T4 gene 32
- Optimized reaction buffer, magnesium chloride, and dNTPs
Specifications
| Lyo-ready T4 UvsX Protein | |
| RPA, RT-RPA | |
| Nucleic Acid Amplification | |
| 42°C | |
| 2 mg/mL |
| Liquid | |
| 120 μg | |
| Lyo-ready T4 UvsX Protein, 60 μL, Store at -25°C to -15°C. | |
| Dry Ice | |
| Up to 20 min. |
For Research Use Only. Not for use in diagnostic procedures.